ENA638586058 Enamine

1-cyclopropyl-2-(3,3-dimethylazetidin-2-yl)-1H-imidazole

  • MDL number MFCD30535619
  • Cas Number : 2060026-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-312101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312101
Availability details for
Quantity
  • Availability
  • Add to Cart