ENA638586084 Enamine

N-{1-cyano-1-[(2-fluorophenyl)methyl]ethyl}acetamide

  • MDL number MFCD30535624
  • Cas Number : 2060044-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-314121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-314121
Availability details for
Quantity
  • Availability
  • Add to Cart