ENA638586097 Enamine

N-(pyridin-3-yl)benzenecarbonimidoyl chloride hydrochloride

  • Cas Number : 2060523-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315838
Availability details for
Quantity
  • Availability
  • Add to Cart