ENA638586105 Enamine

benzyl 4,4-dimethoxy-3-oxopiperidine-1-carboxylate

  • MDL number MFCD30497592
  • Cas Number : 2059938-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-316269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-316269
Availability details for
Quantity
  • Availability
  • Add to Cart