ENA638586109 Enamine

methyl 3-(3-methoxyphenyl)-3-(1H-1,2,3,4-tetrazol-1-yl)propanoate

  • MDL number MFCD30497610
  • Cas Number : 2060031-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317659
Availability details for
Quantity
  • Availability
  • Add to Cart