ENA638586117 Enamine

(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)methanamine hydrochloride

  • MDL number MFCD30497634
  • Cas Number : 2060031-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317850
Availability details for
Quantity
  • Availability
  • Add to Cart