ENA638586151 Enamine

1-ethyl-2-isocyanatobenzene

  • MDL number MFCD00002013
  • Cas Number : 40411-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 79
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147125

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P305+P351+P338,P312,P330,P332+P313,P337+P313,P342+P311,P362,P363,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-147125
s_phrases_eu 23-26-28-37/39
pictograms GHS07,GHS08
hstatements H302,H312,H315,H319,H332,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart