ENA638586195 Enamine

3-amino-3-cyclopropylbutanoic acid

  • MDL number MFCD18699222
  • Cas Number : 1270536-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170343
Availability details for
Quantity
  • Availability
  • Add to Cart