ENA638586216 Enamine

1-bromocyclobutane-1-carbonyl chloride

  • MDL number MFCD30164550
  • Cas Number : 70339-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177605

Safety Information

signalword Danger
pcodes P210,P234,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P390,P403+P233,P403+P235,P405,P406,P501
hcodes C,Xi
MSDS https://www.enaminestore.com/catalog/EN300-177605
pictograms GHS05,GHS07
hstatements H227,H290,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart