ENA638586232 Enamine

methyl 8-nitro-3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylate

  • MDL number MFCD30497429
  • Cas Number : 1403772-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181932
Availability details for
Quantity
  • Availability
  • Add to Cart