ENA638586251 Enamine

4-(4-chlorobenzoyl)-1H-1,2,3-triazole

  • MDL number MFCD23918565
  • Cas Number : 5990-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190529
Availability details for
Quantity
  • Availability
  • Add to Cart