ENA638586280 Enamine

3,4-dihydro-2H-1-benzothiopyran-4-carbonitrile

  • MDL number MFCD30535576
  • Cas Number : 120783-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198331
Availability details for
Quantity
  • Availability
  • Add to Cart