ENA638586315 Enamine

2-(2-sulfanylpyrimidin-5-yl)ethan-1-ol

  • MDL number MFCD30087972
  • Cas Number : 1889531-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211310
Availability details for
Quantity
  • Availability
  • Add to Cart