ENA638586330 Enamine

4-(chloromethoxy)oxane

  • MDL number MFCD21947483
  • Cas Number : 139084-54-1
  • Molecular Weight : 150.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-215319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215319
Availability details for
Quantity
  • Availability
  • Add to Cart