ENA638586352 Enamine

2-{1-[(tert-butoxy)carbonyl]pyrrolidin-3-yl}-2-fluoroacetic acid

  • MDL number MFCD30497440
  • Cas Number : 1781164-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-223204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223204
Availability details for
Quantity
  • Availability
  • Add to Cart