ENA638586406 Enamine

6-bromo-[1,2,4]triazolo[1,5-a]pyridine-8-carboxylic acid

  • MDL number MFCD30497459
  • Cas Number : 1899934-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-253909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253909
Availability details for
Quantity
  • Availability
  • Add to Cart