ENA638586408 Enamine

4,4,5,5-tetramethyl-2-[2-(trifluoromethyl)thiophen-3-yl]-1,3,2-dioxaborolane

  • MDL number MFCD30226957
  • Cas Number : 2246611-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-254398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-254398
Availability details for
Quantity
  • Availability
  • Add to Cart