ENA638586418 Enamine

1-[(tert-butoxy)carbonyl]-4-(2-methoxyethyl)piperidine-4-carboxylic acid

  • MDL number MFCD27995832
  • Cas Number : 1231158-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-258906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258906
Availability details for
Quantity
  • Availability
  • Add to Cart