ENA638586451 Enamine

2-phenylprop-2-en-1-ol

  • MDL number MFCD18827455
  • Cas Number : 6006-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-264486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264486
Availability details for
Quantity
  • Availability
  • Add to Cart