ENA638586452 Enamine

2-methyl-1H-imidazo[4,5-c]pyridine-7-carboxylic acid

  • MDL number MFCD30497475
  • Cas Number : 1783316-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-264595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264595
Availability details for
Quantity
  • Availability
  • Add to Cart