ENA638586459 Enamine

tert-butyl 2-hydrazinylpyridine-3-carboxylate

  • MDL number MFCD28388624
  • Cas Number : 1374849-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-267099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267099
Availability details for
Quantity
  • Availability
  • Add to Cart