ENA638586461 Enamine

3,3-difluoro-2,2-dimethylpentan-1-amine

  • MDL number MFCD30497485
  • Cas Number : 1895193-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-268160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268160
Availability details for
Quantity
  • Availability
  • Add to Cart