ENA638586462 Enamine

3,3-difluoro-2,2-dimethylpentanal

  • MDL number MFCD30497486
  • Cas Number : 2060030-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-268164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268164
Availability details for
Quantity
  • Availability
  • Add to Cart