ENA638586474 Enamine

1,1-dioxo-2,3-dihydro-1lambda6-benzothiophene-2-carboxylic acid

  • MDL number MFCD30535592
  • Cas Number : 1895985-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-270048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-270048
Availability details for
Quantity
  • Availability
  • Add to Cart