ENA638586527 Enamine

4-bromo-5-fluoro-2-methylbenzene-1-sulfonamide

  • MDL number MFCD12026238
  • Cas Number : 1208076-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-286311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-286311
Availability details for
Quantity
  • Availability
  • Add to Cart