ENA638586528 Enamine

2,3,3-trimethylbutane-1-sulfonyl chloride

  • MDL number MFCD26622190
  • Cas Number : 1540450-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-291413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-291413
Availability details for
Quantity
  • Availability
  • Add to Cart