ENA638586529 Enamine

1-methyl-4-nitro-1H-pyrazole-3-sulfonyl chloride

  • MDL number MFCD30164446
  • Cas Number : 1697720-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-291900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-291900
Availability details for
Quantity
  • Availability
  • Add to Cart