ENA638586552 Enamine

(1S,3r,5R)-spiro[bicyclo[3.3.1]nonane-3,2'-oxirane]-7-one

  • MDL number MFCD30535597
  • Cas Number : 53110-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-300366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300366
Availability details for
Quantity
  • Availability
  • Add to Cart