ENA638586571 Enamine

2-(2-nitrophenyl)-1,3-oxazole

  • MDL number MFCD15527551
  • Cas Number : 62882-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-303551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-303551
Availability details for
Quantity
  • Availability
  • Add to Cart