ENA638586598 Enamine

1-[2-(pyrrolidin-1-yl)ethyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12139622
  • Cas Number : 1155065-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-305587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305587
Availability details for
Quantity
  • Availability
  • Add to Cart