ENA638586623 Enamine

5-(benzyloxy)-3H-spiro[1-benzofuran-2,1'-cyclopropane]-3-one

  • MDL number MFCD30535610
  • Cas Number : 72492-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-307622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-307622
Availability details for
Quantity
  • Availability
  • Add to Cart