ENA638586641 Enamine

6-(2-chlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD16069789
  • Cas Number : 147283-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310207
Availability details for
Quantity
  • Availability
  • Add to Cart