ENA638586650 Enamine

ethyl 2-[4-methyl-5-(methylamino)-4H-1,2,4-triazol-3-yl]acetate

  • MDL number MFCD26052668
  • Cas Number : 1567078-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310475
Availability details for
Quantity
  • Availability
  • Add to Cart