ENA638586661 Enamine

1,4-dioxa-10-azadispiro[4.2.4^{8}.2^{5}]tetradecan-11-one

  • MDL number MFCD24566289
  • Cas Number : 914780-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310965
Availability details for
Quantity
  • Availability
  • Add to Cart