ENA638586726 Enamine

1-(ethylsulfanyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD21127243
  • Cas Number : 1490361-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-312763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312763
Availability details for
Quantity
  • Availability
  • Add to Cart