ENA638586749 Enamine

1-(3,5-dimethylphenyl)-2,5-dimethyl-1H-pyrrole

  • MDL number MFCD03576686
  • Cas Number : 119516-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-313640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-313640
Availability details for
Quantity
  • Availability
  • Add to Cart