ENA638586779 Enamine

methyl 4-amino-3-[(methylcarbamoyl)methoxy]benzoate

  • MDL number MFCD12410241
  • Cas Number : 1096309-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-314484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-314484
Availability details for
Quantity
  • Availability
  • Add to Cart