ENA638586780 Enamine

4-amino-3-[(methylcarbamoyl)methoxy]benzoic acid

  • MDL number MFCD12168724
  • Cas Number : 1096900-75-2
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-314485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-314485
Availability details for
Quantity
  • Availability
  • Add to Cart