ENA638586807 Enamine

5-(bromomethyl)-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD30497582
  • Cas Number : 245095-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315370
Availability details for
Quantity
  • Availability
  • Add to Cart