ENA638586815 Enamine

(pentan-3-yl)thiourea

  • MDL number MFCD10692845
  • Cas Number : 854654-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315522
Availability details for
Quantity
  • Availability
  • Add to Cart