ENA638586820 Enamine

(2R)-2-(dimethylamino)butanoic acid

  • MDL number MFCD30497585
  • Cas Number : 2059912-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315725
Availability details for
Quantity
  • Availability
  • Add to Cart