ENA638586827 Enamine

5-bromo-N'-hydroxypyridine-2-carboximidamide

  • Cas Number : 380380-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315793
Availability details for
Quantity
  • Availability
  • Add to Cart