ENA638586844 Enamine

[2-(2-methoxypyridin-4-yl)ethyl](methyl)amine

  • MDL number MFCD28339849
  • Cas Number : 1514115-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-316115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-316115
Availability details for
Quantity
  • Availability
  • Add to Cart