ENA638586912 Enamine

6-oxo-1,6-dihydropyridazine-4-carbonitrile

  • MDL number MFCD22491591
  • Cas Number : 1934386-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317568

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-317568
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart