ENA638586921 Enamine

tert-butyl N-(1-cyclohexyl-1,3-dihydroxypropan-2-yl)carbamate

  • MDL number MFCD30497604
  • Cas Number : 2060006-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317608
Availability details for
Quantity
  • Availability
  • Add to Cart