ENA638586925 Enamine

1-benzyl-4-(oxan-4-yl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD30497606
  • Cas Number : 2060063-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317640
Availability details for
Quantity
  • Availability
  • Add to Cart