ENA638586947 Enamine

1,7,7-trimethyl-1,4-diazepane

  • MDL number MFCD30535640
  • Cas Number : 933716-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317731
Availability details for
Quantity
  • Availability
  • Add to Cart