ENA638586953 Enamine

2-(3-ethyloxetan-3-yl)acetonitrile

  • MDL number MFCD28654220
  • Cas Number : 1783497-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317771
Availability details for
Quantity
  • Availability
  • Add to Cart