ENA638586990 Enamine

[2,5-dimethyl-1-(3-methylphenyl)-1H-pyrrol-3-yl]methanamine hydrochloride

  • MDL number MFCD30497635
  • Cas Number : 2059932-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317852
Availability details for
Quantity
  • Availability
  • Add to Cart