ENA638587221 Enamine

{[(2-cyanophenyl)methyl]sulfanyl}methanimidamide hydrochloride

  • MDL number MFCD30497670
  • Cas Number : 112941-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-318357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-318357
Availability details for
Quantity
  • Availability
  • Add to Cart